missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 5-chloro-2-indolecarboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
Specifica
Melting Point | 166°C to 171°C |
Color | Brown or Tan to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 5 g |
Formula Weight | 223.66 |
Percent Purity | 97% |
Physical Form | Crystals or Powder |
Chemical Name or Material | Ethyl 5-chloro-2-indolecarboxylate, 97% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'