missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetoacetate 99%, ACROS Organics™

Brand:  Acros Organics 117970010

27.00 EUR valid until 2018-12-31
Use promo code "13295" to get your promotional price.



Code : 29

Additional Details : CAS Number : 141-97-9 Weight : 1.07100kg

Packaging Glass Bottle
Quantity 1L

 View more versions of this product

Product Code. 10429030

  • 30.50 € / 1 L


EU Stock 16
Add to basket

Specifications

Chemical Identifiers

141-97-9
130.143
XYIBRDXRRQCHLP-UHFFFAOYSA-N
8868
C6H10O3
MFCD00009199
Acetoacetic ester, EAA
CHEBI:4893

Specifications

Ethyl acetoacetate
70°C
Colorless
1L
141-97-9
Glass Bottle
1.03
99%
0.1% max. (K.F.)
CH3COCH2CO2C2H5
03, 632
15, 3812
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl 3-oxobutanoate
8868
130.14
1.0300g/mL
180.0°C
-45.0°C
99%
Authentic
1.4180 to 1.4200
Pure
99%
C6H10O3
MFCD00009199
12,222
Acetoacetic ester, EAA
CCOC(=O)CC(=O)C
130.143
CHEBI:4893
Liquid
Videos
SDS Special Offers

Special Offers

  • Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    : 13295

    2018-12-31

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/09/2018 and 31/12/2018. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions