missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Alcohol, HPLC/Spectrophotometric Grade, Pure, Honeywell™
Click to view available options
Quantity:
1 L
2 L
Specifications
Linear Formula | CH3CH2OH |
UN Number | UN1170 |
Formula Weight | 46.07g/mol |
Grade | HPLC/Spectrophotometric |
Concentration or Composition (by Analyte or Components) | 200 proof |
Density | 0.789g/mL at 20°C |
Quantity | 2 L |
Flash Point | 14°C (57.2°F) |
Chemical Name or Material | Ethyl alcohol, Pure |
Safety and Handling
Product Identifier
- Ethyl alcohol, Pure
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : ethanol
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'