missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl butyrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acidity | 1mg KOH/g max. |
Melting Point | -93.3°C |
Color | Colorless |
Density | 0.8700g/mL |
Boiling Point | 120.0°C |
Flash Point | 26°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH3CH2CH2CO2C2H5 |
Show More |
Safety and Handling
Product Identifier
- Ethyl butyrate
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P403+P235-Store in a well-ventilated place. Keep cool.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'