missing translation for 'onlineSavingsMsg'
Learn More

Ethyl ethynyl ether, 40 wt% solution in hexanes, Thermo Scientific™

Brand:  Thermo Scientific Acros 307390010

Code : 29

Additional Details : CAS Number : 927-80-0 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 3 Pack group : II

Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 10037782

  • 37.30 € / 1g
In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

927-80-0
70.09
WMYNMYVRWWCRPS-UHFFFAOYSA-N
61239
CCOC#C
C4H6O
MFCD00009247
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
ethynoxyethane

Specifications

Ethyl ethynyl ether
0.7180g/mL
55.0°C to 56.0°C
confirmed by 1H-NMR
C2H5OC≡CH
ca. 40 wt% (typically 35 to 55 wt%)
ethoxyacetylene, ethoxyethyne, ethyl ethynyl ether, ethyne, ethoxy, ether, ethyl ethynyl, ethoxy-ethyne, ethyne, ethoxy-9ci, 1-ethoxyacetylene, ethoxyacetylene w/w solution in hexanes, ethoxy acetylene
WMYNMYVRWWCRPS-UHFFFAOYSA-N
CCOC#C
70.09
70.09
927-80-0,92112-69-1
Orange to Brown
−28°C
C4H6O
MFCD00009247
Glass bottle
1g
0.718
ethynoxyethane
61239
Liquid
Videos
SDS missing translation for 'documents'
Product Certifications
Special Offers

Special Offers

RUO – Research Use Only