missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isonipecotate, 98+%, Thermo Scientific Chemicals

Catalog Number p-130368
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1126-09-6
C8H15NO2
157.21
MFCD00006003
RUJPPJYDHHAEEK-UHFFFAOYSA-N
ethyl 4-piperidinecarboxylate, ethyl isonipecotate, piperidine-4-carboxylic acid ethyl ester, ethylpiperidine-4-carboxylate, 4-piperidinecarboxylic acid, ethyl ester, isonipecotic acid ethyl ester, 4-piperidinecarboxylic acid ethyl ester, 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine
70770
ethyl piperidine-4-carboxylate
CCOC(=O)C1CCNCC1
This item is not returnable. View return policy
1126-09-6
C8H15NO2
157.21
MFCD00006003
RUJPPJYDHHAEEK-UHFFFAOYSA-N
ethyl 4-piperidinecarboxylate, ethyl isonipecotate, piperidine-4-carboxylic acid ethyl ester, ethylpiperidine-4-carboxylate, 4-piperidinecarboxylic acid, ethyl ester, isonipecotic acid ethyl ester, 4-piperidinecarboxylic acid ethyl ester, 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine
70770
ethyl piperidine-4-carboxylate
CCOC(=O)C1CCNCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown to Colorless
Density 1.0200g/mL
Boiling Point 95.0°C to 98.0°C (7.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4585 to 1.4605
Quantity 25 g
Beilstein 22, IV, 128
Specific Gravity 1.02
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 157.21
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Ethyl isonipecotate, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl isonipecotate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'