missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl maltol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 89.0°C to 93.0°C |
Boiling Point | 289.0°C to 290.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 100 g |
Merck Index | 15,3879 |
Synonym | 3-benzyloxy cyclobutanecarboxylic acid, 3-benzyloxy-cyclobutanecarboxylic acid, 3-benzyloxy cyclobutane-1-carboxylic acid, 3-benzyloxycyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-phenylmethoxy, trans-3-benzyloxycyclobutanecarboxylic acid, cis-3-benzyloxy cyclobutanecarboxylic acid, trans-3-benzyloxy cyclobutanecarboxylic acid, cis-3-benzyloxycyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 3-phenylmethoxy-, trans |
InChI Key | YNNOFVDQHAHVFG-UHFFFAOYSA-N |
SMILES | C1C(CC1OCC2=CC=CC=C2)C(=O)O |
IUPAC Name | 3-phenylmethoxycyclobutane-1-carboxylic acid |
Show More |
Safety and Handling
Product Identifier
- Ethyl maltol, 98%
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'