missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-BOC-4-piperidinecarboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-4228649
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
142851-03-4
C13H23NO4
257.33
MFCD01763998
MYHJCTUTPIKNAT-UHFFFAOYSA-N
ethyl n-boc-piperidine-4-carboxylate, 1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate, ethyl n-boc-4-piperidinecarboxylate, n-boc-4-carbethoxy piperidine, 1,4-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 4-ethyl ester, ethyl n-boc-isonipecotate, ethyl 1-boc-piperidine-4-carboxylate, dimethylethyl piperidinedicarboxylic acid ethyl ester, 1-boc-piperidine-4-carboxylic acid ethyl ester, 1-boc-isonipecotic acid ethyl ester
2758812
1-O-tert-butyl 4-O-ethyl piperidine-1,4-dicarboxylate
CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
142851-03-4
C13H23NO4
257.33
MFCD01763998
MYHJCTUTPIKNAT-UHFFFAOYSA-N
ethyl n-boc-piperidine-4-carboxylate, 1-tert-butyl 4-ethyl piperidine-1,4-dicarboxylate, ethyl n-boc-4-piperidinecarboxylate, n-boc-4-carbethoxy piperidine, 1,4-piperidinedicarboxylic acid, 1-1,1-dimethylethyl 4-ethyl ester, ethyl n-boc-isonipecotate, ethyl 1-boc-piperidine-4-carboxylate, dimethylethyl piperidinedicarboxylic acid ethyl ester, 1-boc-piperidine-4-carboxylic acid ethyl ester, 1-boc-isonipecotic acid ethyl ester
2758812
1-O-tert-butyl 4-O-ethyl piperidine-1,4-dicarboxylate
CCOC(=O)C1CCN(CC1)C(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0460g/mL
Boiling Point 120.0°C to 135.0°C (0.5 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.046
Formula Weight 257.33
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl N-BOC-4-piperidinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl N-Boc-4-piperidinecarboxylate, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'