missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl phenylglyoxylate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.
Solubility
Soluble in water 1143 mg/L @ 25°C .
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Ethyl phenylglyoxylate has been used in preparation of 1,5-dihydro-5-deazaflavin derivatives possessing a chiral substituent at N(3) position.
Solubility
Soluble in water 1143 mg/L @ 25°C .
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Specifications
Density | 1.122 |
Boiling Point | 138°C to 139°C (18 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.516 |
Quantity | 5 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water 1143mg/L @ 25°C . |
Formula Weight | 178.19 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl phenylglyoxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'