missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-4-cyano-3-hydroxybutanoate, 95%, Thermo Scientific™

Catalog Number 11935621
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
141942-85-0
C7H11NO3
157.17
MFCD00270839
LOQFROBMBSKWQY-ZCFIWIBFSA-N
ethyl r---4-cyano-3-hydroxybutyate, ethyl r---4-cyano-3-hydroxybutyrate, ethyl 3r-4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxybutyric acid ethyl ester, r-ethyl 4-cyano-3-hydroxybutyrate, ethyl-r-+-4-cyano-3-hydroxybutanoate, ethyl r-4-cyano-3-hydroxybutyrate, r-ethyl 4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxy-butyric acid ethyl ester, r---4-cyano-3-hydroxybutyric acid ethyl ester
2733879
ethyl (3R)-4-cyano-3-hydroxybutanoate
CCOC(=O)C[C@H](O)CC#N
This item is not returnable. View return policy
141942-85-0
C7H11NO3
157.17
MFCD00270839
LOQFROBMBSKWQY-ZCFIWIBFSA-N
ethyl r---4-cyano-3-hydroxybutyate, ethyl r---4-cyano-3-hydroxybutyrate, ethyl 3r-4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxybutyric acid ethyl ester, r-ethyl 4-cyano-3-hydroxybutyrate, ethyl-r-+-4-cyano-3-hydroxybutanoate, ethyl r-4-cyano-3-hydroxybutyrate, r-ethyl 4-cyano-3-hydroxybutanoate, r-4-cyano-3-hydroxy-butyric acid ethyl ester, r---4-cyano-3-hydroxybutyric acid ethyl ester
2733879
ethyl (3R)-4-cyano-3-hydroxybutanoate
CCOC(=O)C[C@H](O)CC#N
Color Colorless to Yellow
Density 1.1140g/mL
Boiling Point 270.0°C
Flash Point >190°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula NCCH2CH(OH)CH2COOCH2CH3
Refractive Index 1.4460 to 1.4500
Quantity 1 g
Specific Gravity 1.114
Specific Rotation -4.8° to -5.2° (20°C, 589nm) (neat)
Solubility Information Solubility in water: soluble in water
Formula Weight 157.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl (R)-4-cyano-3-hydroxybutanoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl (R)-4-cyano-3-hydroxybutanoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'