missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tetrahydropyran-4-ylacetate, 97%, Thermo Scientific™

Catalog Number p-4391344
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
103260-44-2
C9H16O3
172.224
JLMMMEDWRUVCEW-UHFFFAOYSA-N
ethyl tetrahydropyran-4-ylacetate, ethyl 2-tetrahydro-2h-pyran-4-yl acetate, ethyl 2-oxan-4-yl acetate, ethyl tetrahydropyran-4-yl-acetate, ethyl 2-4-tetrahydropyranyl acetate, ethyl tetrahydropyranyl-4-acetate, 2h-pyran-4-acetic acid, tetrahydro-, ethyl ester, ethyl tetrahydropyran-4-acetate, tetrahydro-pyran-4-yl-acetic acid ethyl ester, ethyl tetrahydro-2h-pyran-4-ylacetate
2773412
ethyl 2-(oxan-4-yl)acetate
CCOC(=O)CC1CCOCC1
This item is not returnable. View return policy
103260-44-2
C9H16O3
172.224
JLMMMEDWRUVCEW-UHFFFAOYSA-N
ethyl tetrahydropyran-4-ylacetate, ethyl 2-tetrahydro-2h-pyran-4-yl acetate, ethyl 2-oxan-4-yl acetate, ethyl tetrahydropyran-4-yl-acetate, ethyl 2-4-tetrahydropyranyl acetate, ethyl tetrahydropyranyl-4-acetate, 2h-pyran-4-acetic acid, tetrahydro-, ethyl ester, ethyl tetrahydropyran-4-acetate, tetrahydro-pyran-4-yl-acetic acid ethyl ester, ethyl tetrahydro-2h-pyran-4-ylacetate
2773412
ethyl 2-(oxan-4-yl)acetate
CCOC(=O)CC1CCOCC1
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 172.22
Percent Purity 97%
Chemical Name or Material Ethyl tetrahydropyran-4-ylacetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'