missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha-bromophenylacetate, 97%, ACROS Organics™

Brand:  Acros Organics 438061000

Code : 29

Additional Details : CAS Number : 2882-19-1 Weight : 0.15000kg Transport : UN number : 3265 Chem class : 8 Pack group : II


Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 11955001

  • 319.00 € / 100g

Estimated Shipment
26-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2882-19-1
243.1
ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate
ethyl 2-bromo-2-phenylacetate
C10H11BrO2
BKTKLDMYHTUESO-UHFFFAOYSA-N
97780
CCOC(=O)C(C1=CC=CC=C1)Br

Specifications

Ethyl α-bromophenylacetate
>110°C
Glass bottle
1.389
Colorless to Yellow
2882-19-1
C10H11BrO2
ethyl alpha-bromophenylacetate, ethyl bromophenylacetate, ethyl bromo phenyl acetate, ethyl-alpha-bromophenyl acetate, ethyl-.alpha.-bromophenyl acetate, benzeneacetic acid, .alpha.-bromo-, ethyl ester, bromo-phenyl-acetic acid ethyl ester, benzeneacetic acid, alpha-bromo-, ethyl ester, eyr&vo2, ethylbromo phenyl acetate
CCOC(=O)C(C1=CC=CC=C1)Br
243.1
243.1
97%
1.3890g/mL
Authentic
1.5385 to 1.5405 (20°C, 589nm)
89°C to 92°C (1.2 hPa)
100g
96% min. (GC)
C6H5Ch(Br)COOCH2CH3
BKTKLDMYHTUESO-UHFFFAOYSA-N
ethyl 2-bromo-2-phenylacetate
97780
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl alpha-bromophenylacetate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents