Learn More
ethyle3,5-dichlorobenzoate, 97%, Maybridge™
Brand: Maybridge TL00759DA
Code : 31
Additional Details : CAS Number : 91085-56-2 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
91085-56-2 | |
219.061 | |
JRLNLVFPSMDPLU-UHFFFAOYSA-N | |
2736404 | |
CCOC(=O)C1=CC(=CC(=C1)Cl)Cl |
C9H8Cl2O2 | |
MFCD00221467 | |
ethyl3,5-dichlorobenzoate, 3,5-dichlorobenzoic acid ethyl ester, rarechem al bi 0057, 5-ethoxycarbonyl-1,3-dichlorobenzene, benzoic acid, 3,5-dichloro-, ethyl ester, pubchem10323, ethyl 3,5-dichloro benzoate, ethyl 3,5-bis chloranyl benzoate, 3,5-dichlorobenzoic acid ethyl etster | |
ethyl 3,5-dichlorobenzoate |
Specifications
Ethyl 3, 5-dichlorobenzoate | |
1g | |
97.0 | |
C9H8Cl2O2 | |
MFCD00221467 | |
JRLNLVFPSMDPLU-UHFFFAOYSA-N | |
ethyl 3,5-dichlorobenzoate | |
2736404 | |
97% |
Amber Glass Bottle | |
91085-56-2 | |
97% | |
Cl2C6H3C(O)OCH2CH3 | |
ethyl3,5-dichlorobenzoate, 3,5-dichlorobenzoic acid ethyl ester, rarechem al bi 0057, 5-ethoxycarbonyl-1,3-dichlorobenzene, benzoic acid, 3,5-dichloro-, ethyl ester, pubchem10323, ethyl 3,5-dichloro benzoate, ethyl 3,5-bis chloranyl benzoate, 3,5-dichlorobenzoic acid ethyl etster | |
CCOC(=O)C1=CC(=CC(=C1)Cl)Cl | |
219.061 | |
219.07 |