missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical™

Catalog Number p-7073124
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Bag in box
HDPE plastic bottle
139-33-3
C10H14N2Na2O8
336.21
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ZGTMUACCHSMWAC-UHFFFAOYSA-L
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
57339238
CHEBI:64734
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
139-33-3
C10H14N2Na2O8
336.21
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ZGTMUACCHSMWAC-UHFFFAOYSA-L
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
57339238
CHEBI:64734
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP

TRUSTED_SUSTAINABILITY
Color Colorless
Packaging HDPE plastic bottle
Quantity 2.5 L
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Physical Form Liquid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'