missing translation for 'onlineSavingsMsg'
Learn More

Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 10427880
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
4755-77-5
C4H5ClO3
136.53
MFCD00000706
OWZFULPEVHKEKS-UHFFFAOYSA-N
ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
20884
ethyl 2-chloro-2-oxoacetate
CCOC(=O)C(=O)Cl
This item is not returnable. View return policy
4755-77-5
C4H5ClO3
136.53
MFCD00000706
OWZFULPEVHKEKS-UHFFFAOYSA-N
ethyl chlorooxoacetate, ethyl oxalyl chloride, ethyl chloroglyoxylate, ethoxalyl chloride, ethyl oxalyl monochloride, acetic acid, chlorooxo-, ethyl ester, oxalic acid monoethyl ester chloride, monoethyl oxaloyl chloride, oxalyl chloride, ethyl ester, ethyl chlorocarbonyl formate
20884
ethyl 2-chloro-2-oxoacetate
CCOC(=O)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Density 1.2200g/mL
Boiling Point 135°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCO2C2H5
Refractive Index 1.4160 to 1.418
Quantity 25 mL
Beilstein 02, 541
Specific Gravity 1.22
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in common organic solvents
Formula Weight 136.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl oxalyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl oxalyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'