missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Eutech™ PC 450 Multi-parameter Meter Kit
Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
695.00 € - 978.00 €
Specifications
Outputs | USB or RS232 |
---|---|
AC Adapter | Optional universal power adapter |
Accuracy (Conductivity) | ± 1% full scale |
Accuracy (mV) | ±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater |
Accuracy (pH) | ±0.01 |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15327063
|
Thermo Scientific™
01X651636 |
PC 450 Meter Only: Meter, two AA batteries |
695.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15357063
|
Thermo Scientific™
01X651637 |
PC 450 Meter & Combi Electrode: Meter, combi pH/Con/ATC electrode, two AA batteries |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Your all-in-one solution for multi-parameter research work in the lab. The multi-parameter Eutech PC 450 measures pH / mV / Conductivity / Total Dissolved Solids / Temperature. It accepts a single pH/Con probe or separate pH and conductivity probes.
Features
- Switch between pH, mV, conductivity and TDS with a press of a button
- Large, backlit custom display – Lets you view measurement with temperature
- Grip-Clip™ – Quickly and easily secures separate pH and conductivity electrodes simultaneously to your sample container or beaker
- Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards you
- Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
- Intuitive, user-friendly icons – Electrode status displays pH slope or cell constant
- Auto-range conductivity from 0 to 199.9 mS in five ranges – With up to five calibration points
- Selectable cell constant – 0.10, 1.00 or 10.0
- Adjustable temperature coefficient – From 0.00 to 10.00 % per ºC
- Conductivity-to-TDS factor – Adjustable from 0.40 to 1.00
- Selectable ºC/ºF – Choose units to best fit your application
- Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
- Store up to 500 data sets; easy USB or RS232 output with optional data cables
- Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
- Built-in stand with wall-mount – Allows for easy benchtop measurement and storage
Specifications
USB or RS232 | |
± 1% full scale | |
±0.01 | |
Calibration due alarm | |
Up to 6 points: USA, NIST, DIN or custom | |
LCD Graphic, Backlit | |
pH, ORP, Ion, Conductivity, TDS, Salinity, Resistivity | |
±2000 mV | |
0 to 200 ppt (TDS factor 1.0) | |
-10.0°C to +110.0°C | |
Two AA batteries (included), >100 hours (without backlight); AC adapter (optional) | |
0.1 mV ±999.9 mV; 1 mV beyond | |
±0.5°C, ±0.9°F | |
0.1 °C/°F | |
Meter: 1 lb |
Optional universal power adapter | |
±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater | |
± 1% full scale | |
Auto/manual; one point per range | |
Temperature: Automatic or manual from -10 to 110 °C | |
IP 67 | |
0 to 200 mS, 0 to 20 mS if using COMBI03BW probe | |
-2.00 to 16.00 | |
0.01 | |
Stores up to 500 data sets | |
0.01 μS to 0.1 mS | |
0.01 ppm to 0.1 ppt | |
14.0°F to 230.0°F | |
Meter: 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm) | |
Meter: 0.5 kg |