Learn More
Thermo Scientific™ Eutech™ PD 450 Meter
Collecting multi-parameter data is a breeze using Eutech™ PD 450 Meter.
Brand: Thermo Scientific™ 01X651646
Code : NEW
Additional Details : Weight : 1.50000kg
Description
The Eutech™ PD450 Meter is a multi-parameter meter that measures pH, mV, dissolved oxygen, and temperature. Affordable optical RDO technology reduces maintenance time and costs. The maintenance free optical probe with no electrolyte eliminates the hassle of replacing delicate membranes and salt build up from refillling solutions. You can also easily switch between pH, mV, % saturation and mg/L (ppm) dissolved oxygen – with the press of a button.
Features:
- pH / mV / Dissolved Oxygen / Temperature
- Durable waterproof and dustproof design with IP67 rating
- Large, backlit graphic display provides measurement with temperature
- Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards
- Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
- Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
- Selectable ºC or ºF – Choose units to best fit your application
- Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
- Store up to 500 data sets – Easy USB or RS232 output with optional data cables
- Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
Specifications
USB or RS232 | |
±2 %; ±0.1 ppm or mg/L up to 8; ±0.2 ppm or mg/L above 8 | |
±0.01 | |
2 AA (included) | |
Up to 6 points: USA, NIST, DIN or custom | |
PD 450 Meter & Electrodes: Meter, pH/ATC electrode, RDO electrode with sensor cap, two AA batteries | |
IP 67 | |
±2000 mV | |
0.01 | |
Time and date stamp on calibration and stored data | |
0 to 200 %; 0 to 20 ppm/mg/L | |
0.10 %; 0.1 ppm/mg/L | |
Automatic or manual from -10 to 110 °C | |
0.1 ºC/ºF | |
Meter: 1 lb , Boxed: 1.4 lb |
Optional | |
±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater | |
100+ hr. (without backlight) | |
mV: ±200 mV, DO: Zero and saturation, Temp: Offset in 0.1 increments, ±10 °C or & | |
Pressure & Salinity Compensation: manual | |
Backlit Custom LCD | |
pH, mV, Dissolved Oxygen, Temperature | |
-2.00 to 16.00 | |
-10.0°C to +110.0°C | |
Stores up to 500 data sets | |
0.1 mV ±999.9 mV; 1 mV beyond | |
±0.5°C, ±0.9°F | |
14.0°F to 230.0°F | |
Meter: 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm), Boxed: 9 1/8in x 9 1/8in x 2 3/4in (23 x 23 x 7 cm) | |
Meter: 0.5 kg, Boxed: 0.7 kg |