Learn More
Thermo Scientific™ Eutech™ pH 700 Meter
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools.
Brand: Thermo Scientific™ 01X541610
Code : 04
Additional Details : Weight : 1.00000kg
Description
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools. It features a large, easy-to-read display, ready indicator that letsyou know when readings are stable and up to 5-point calibration with auto-buffer recognition. It also has a non-volatile memory holds up to 100 data points and comes witu an integral electrode holder.
Features
- Large, easy-to-read display
- Ready indicator let you know when readings are stable
- Up to 5-point calibration with auto-buffer recognition
- Non-volatile memory holds up to 100 data points
- Integral electrode holder
Applications
- Educational: Useful for most laboratory, ecological studies and other applications.
- Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing. Use in all types of food processing.
Specifications
Product Specification: Description
pH
- Range: -2.00 to 16.00 pH
- Resolution: 0.01 pH
- Accuracy: ±0.01 pH
- Cal. Points: Up to 5
- Buffer Sets: USA (1.68, 4.01, 7.00, 10.01, 12.45), NIST (1.68, 4.01, 6.86, 9.18, 12.45)
ORP
- Range: ±2000 mV
- Rel. mV Range: ±2000 mV
- Resolution: 0.1 mV (±199.9 mV) / 1 mV (beyond)
- Accuracy: ±0.2 mV (±199.9 mV) / ±2 mV (beyond)
Temperature
- Range (Meter): 0.0 to 100.0 ºC / 32.0 to 212.0 ºF
- Resolution: 0.1 ºC / 0.1 ºF
- Accuracy: ±0.3 ºC / ±0.5 ºF (0 to 70 ºC)
- Compensation: ATC / MTC (0 to 100 ºC) (pH only)
- Calibration: Offset in 0.1 ºincrements; offset range: ±5.0 ºC / ±9.0 ºF
- Power Requirements: 9 V DC adapter, 1.3 A (100/240 VAC, SMPS)
Specifications
Eutech pH 700 Meter With Glass Body, Open Pore pH Electrode (EC620131), ATC Probe (PH5TEMB01P) Integral Electrode Holder & 100/240 VAC Adapter | |
±0.2 (±199.9), ±2 (Beyond) | |
Offset: 0.1° increments; Offset Range: ±5.0 °C (±9.0 °F) | |
0.01 | |
±2000 mV | |
0°C to 100°C | |
0.1°C/°F | |
For ORP: ±0.2 mV (±199.9 mV) / ±2 mV (beyond) | |
ATC/MTC (0°C to 100°C) (pH only) |
±0.01 | |
±0.3°C/±0.5°F | |
-2 to 16 | |
±2000 | |
Up to 5 | |
32.0°F to 212.0°F | |
100 | |
For ORP: 0.1 mV (±199.9 mV) / 1 mV (beyond) | |
9 V DC adapter, 1.3 A (100/240 VAC, SMPS) |