Learn More
Thermo Scientific™ Eutech™ Replacement Double Junction Sensor for ORPTestr 10 Meter
Eutech offers a wide range of ORP meters that measure both pH and ORP values in various educational, laboratory and industrial applications.
Brand: Thermo Scientific™ 01X106711
Code : 04
Additional Details : Weight : 1.00000kg
Description
Eutech offers a wide range of ORP meters that measure both pH and ORP values in various educational, laboratory and industrial applications. General appications include chromate reduction, cyanide oxidation, pulp bleaching in industrial settings as well as monitor ozone systems, check free chlorine activity (in pools, spas, and cooling waters), water purification and drinking water, aquarium monitoring, and water pollution monitoring
Features:
- Replacement part for ORP Testr 10 Cat # ORPTEST10
Specifications
Replacement Part | |
ORPTestr 10 Meter |