Learn More
SCAT Exhaust Filter, V3.0
The SCAT Exhaust Filter V3.0 for SafetyWasteCaps guarantees over 90% CTC adsorption for maximum air purity due to three integrated activated carbon types. This makes it the most powerful activated carbon filter medium on the HPLC laboratory market.
Brand:  SCAT 407982
92.46 EUR valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.12900kg
 View more versions of this product
Product Code. 17218870
  • 117.00 € / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Block off harmful vapours and ensure for safe pressure equalization in the waste containers
  • 3-layer filter media with high performance spectrum for acids and alkalis
  • Optimized for binding HPLC buffer solutions and absorbing organic solvent vapors
  • The natural pore structure of the activated carbon prevents sticking or clogging
  • Precise indication of the next replacement time by means of a time indicator
  • Available in sizes (S/M/L) with different service lives (3/6/12 months)
  • The use of exhaust filters can contribute to the reduction of the air exchange rate
Specifications
Specifications
Single Pk. (Service Life 6 Months)
HPLC Systems
Exhaust Filter
SCAT Safety Waste Caps & Universal Waste Hub JAN
Label
M
Polypropylene
GL14