Learn More
SCAT Exhaust Filter, V3.0
The SCAT Exhaust Filter V3.0 for SafetyWasteCaps guarantees over 90% CTC adsorption for maximum air purity due to three integrated activated carbon types. This makes it the most powerful activated carbon filter medium on the HPLC laboratory market.
Brand:  SCAT 407986
134.16 EUR valid until 2024-12-31
Use promo code "23290" to get your promotional price.
Additional Details : Weight : 0.21900kg
 View more versions of this product
Product Code. 17228870
  • 170.00 € / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Block off harmful vapours and ensure for safe pressure equalization in the waste containers
  • 3-layer filter media with high performance spectrum for acids and alkalis
  • Optimized for binding HPLC buffer solutions and absorbing organic solvent vapors
  • The natural pore structure of the activated carbon prevents sticking or clogging
  • Precise indication of the next replacement time by means of a time indicator
  • Available in sizes (S/M/L) with different service lives (3/6/12 months)
  • The use of exhaust filters can contribute to the reduction of the air exchange rate
Specifications
Specifications
Single Pk. (Service Life 12 Months)
HPLC Systems
Exhaust Filter
SCAT Safety Waste Caps & Universal Waste Hub JAN
Indicator
L
Polypropylene
GL14