missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antigen Rat, FITC, Clone: BM8.1, enQuireBio™
Rat Monoclonal Antibody
82.15 € - 360.00 €
Specifications
Antigen | F4/80 Antigen |
---|---|
Clone | BM8.1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942358
|
EnquireBio
QAB68-F-25UG |
25 μg |
82.15 €
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932358
|
EnquireBio
QAB68-F-100UG |
100 μg |
252.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952358
|
EnquireBio
QAB68-F-500UG |
500 μg |
360.00 €
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
F4/80 Antigen | |
0.5mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
13733 | |
IgG2b κ | |
Protein A/G Purified |
BM8.1 | |
Flow Cytometry | |
FITC | |
RUO | |
XP_006523664 | |
Adgre1 | |
Primary | |
Store at 2-4°C. |