missing translation for 'onlineSavingsMsg'
Learn More

Factor XIIIa Mouse, Clone: F13A1/1683, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 2162-MSM3-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15989977

  • 459.10 € / 100µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Factor XIIIa
Cytoplasmic and Secreted
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
F13A1
Recombinant fragment of human Factor XIIIa protein (aa46-181)
83kDa
Protein A/G Purified
RUO
Primary
2162
Immunohistochemistry
F13A1/1683
Unconjugated
NP_000120
Mouse
IgG2b kappa
293T Cells. Placenta.
100 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
SDS
Documents
Certificates