missing translation for 'onlineSavingsMsg'
Learn More

Logistik Unicorp™ Faithful White Ladies' Fitted Lab Coats

Catalog Number 13152190
Click to view available options
Size (Metric) Chest:
120cm
124cm
136cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Use Logistik Unicorp™ Faithful White Ladies' Fitted Lab Coats for general laboratory safety. It features two lower patch pockets, breast pocket, concealed stud-fastened front and stud-fastened cuffs

  • Contoured fit ladies' coat;
  • Two lower patch pockets
  • Breast pocket
  • Concealed stud-fastened front
  • Stud-fastened cuffs
  • Not CE marked
Type Lab Coat
Material Polycotton
Color White
Size (Metric) Chest 124cm
For Use With (Application) Used for general laboratory safety
Includes Two lower patch pockets, breast pocket, concealed stud fastened front, stud fastened cuffs
Weight (Metric) 245 g/m2
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'