missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Green FCF, pure, certified

Catalog Number p-4259092
73.45 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2353-45-9
C37H34N2Na2O10S3
808.84
MFCD00013053
XJBPDZVCYTYRIN-UHFFFAOYSA-L
C.I. 42053
16887
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
This item is not returnable. View return policy
2353-45-9
C37H34N2Na2O10S3
808.84
MFCD00013053
XJBPDZVCYTYRIN-UHFFFAOYSA-L
C.I. 42053
16887
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 290.0°C
Absorption Ratio 0.98 to 1.2
Dye Content 85% min.
Solubility Information Solubility in water: 150g/L (25°C).
Infrared Spectrum Authentic
Lambda Max 622 to 626nm
Formula Weight 808.84
Grade Pure
Packaging Glass bottle
Color Bordeaux to Maroon
Quantity 25 g
Physical Form Powder
Chemical Name or Material Fast Green FCF
Show More Show Less
compliance-icons
Product Identifier
  • Fast green FCF
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'