Learn More
Fistreem International™ Pre-Treat Feed Accessory Kit
Suitable pre-treatment feed kit for the Calypso Water Still. Fistreem International™ Pre-Treat Feed Accessory Kit allows you to connect a suitable pre-treated water supply ( deionized, reverse osmosis or already distilled water ) to your still.
Brand:  Fistreem International™ WCA901.SVA.7
Code : 11
Additional Details : Weight : 5.00000kg
Product Code. 13186020
  • 138.18 € / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Allows you to connect a suitable pre-treated water supply to your still.
  • In the pre-treated mode, the already purified water is fed directly to the boiler and tap water is fed to the condenser coil for cooling purposes only.
Specifications
Specifications
Calypso Water Still
Pre-treat Feed Accessory Kit