Learn More
Thermo Scientific™ Long PCR Enzyme Mix Not available in the USA
A unique enzyme blend for long and tricky PCR applications.
102.00 € - 404.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10460280
|
Thermo Scientific™
K0181 |
100U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10470280
|
Thermo Scientific™
K0182 |
500U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific Long PCR Enzyme Mix is a unique blend of Taq DNA Polymerase and a thermostable high fidelity DNA polymerase with proofreading activity. The two enzymes synergistically generate long PCR products with greater yield and fidelity than Taq DNA Polymerase alone.
The fidelity of PCR with this enzyme mix is three times higher than with Taq DNA Polymerase. The ratio of enzymes in the Long PCR Enzyme Mix is optimized for generation of very long amplicons: up to 47 kb with viral DNA and up to 21 kb with genomic DNA templates. The specially formulated Long PCR Buffer protects DNA from depurination and nicking during long thermal cycling. The products generated with the Long PCR Enzyme Mix are mostly 3'-dA tailed. Long PCR Enzyme Mix is also used for efficient amplification of GC-rich DNA regions.
The two enzymes synergistically generate long PCR products with greater yield and fidelity than Taq DNA Polymerase alone. The fidelity of PCR with this enzyme mix is three times higher than with Taq DNA Polymerase. The ratio of enzymes in the Long PCR Enzyme Mix is optimized for generation of very long amplicons: up to 47kb with viral DNA and up to 21kb with genomic DNA templates (see Fig. 1). The specially formulated Long PCR Buffer protects DNA from depurination and nicking during long thermal cycling. The products generated with the Long PCR Enzyme Mix are mostly 3'-dA tailed.Long PCR Enzyme Mix is also used for efficient amplification of GC-rich DNA regions (see Fig. 2).- Unique blend of two thermostable DNA polymerases
- Long PCR products:—up to 47kb with viral DNA as template,—up to 21kb with genomic DNA as template
- Ideal for GC-rich templates up to 85%
- Fidelity is three-times higher than with Taq DNA polymerase (see Fig. 1)
- High yield
Recommended for:
Long range PCR; PCR from GC-rich or difficult templates; Generation of template for TA cloning; RT-PCR.
Specifications
Long PCR Enzyme Mix | |
Cloning |
This product is licensed under one U.S. Patent No. 5,436,149 or corresponding foreign patents owned by Takara Bio, Inc.