Learn More
Thermo Scientific™ O’RangeRuler 500 bp DNA Ladder, ready-to-use
Perform precise sizing of PCR products and other double-stranded DNA fragments in agarose or non-denaturing polyacrylamide gels with this DNA step ladder of 500 bp increments.
Brand: Thermo Scientific™ SM0643
Code : 88
Additional Details : Weight : 0.00900kg
Description
O'RangeRuler 500 bp DNA Ladder 500-6000 bp is designed for precise sizing of PCR products and other double-stranded DNA fragments in agarose gels. It is step ladder with 500 bp differences between the DNA fragments. It consists of purified and ligated blunt ended basic unit repeats of 500 bp. Not recommended for DNA quantification.
O'RangeRuler 500 bp DNA Ladder is supplied with 6X Orange DNA Loading Dye for sample DNA.
Highlights
- Step ladder with 500 bp increments
- Sharp bands, easy-to-remember fragment sizes
- Evenly spaced reference bands (given in red)
- Ready-to-use -premixed with 6X Orange Loading Dye for direct loading and room temperature storage
- Supplied with loading dye for sample DNA
Specifications
6X | |
Xylene Cyanol FF, Orange G | |
Yes | |
Agarose Gels | |
O’GeneRuler, O’RangeRuler |
Agarose | |
DNA Ladder | |
500 to 6000 bp | |
Sustainable packaging |
For Research Use Only. Not for use in diagnostic procedures.