missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, 7-Hydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 2070-01

Additional Details : CAS Number : 7782-63-0 Weight : 0.65000kg

Packaging Poly Bottle
Quantity 500g

 View more versions of this product

Product Code. 15547954

  • 131.00 € / 500g

Call for latest local availability
01 885 5854

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

7782-63-0
278.006
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
CHEBI:75836
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
FeH14O11S
SURQXAFEQWPFPV-UHFFFAOYSA-L
62662
iron(2+);sulfate;heptahydrate

Specifications

Ferrous Sulfate
7782-63-0
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[Fe+2]
278.006
CHEBI:75836
Granular
Poly Bottle
500g
7-Hydrate
FeH14O11S
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron(2+);sulfate;heptahydrate
62662
278.02
BAKER ANALYZED™ ACS Reagent
1L = 1.90kg
Videos
Documents