missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, Australia

Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia orgin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.

Brand:  Gibco™ 10100147

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10287262

  • 1024.00 € / 500mL
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sera Category: Secure
  • Origin: Australia
  • Endotoxin level: ≤10EU/mL
  • Hemoglobin level: ≤30mg/dL (levels routinely ≤25mg/dL)

Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Fetal Bovine Serum
Heat-inactivated
Sustainable packaging
Qualified
Frozen
Fetal
Australia
Bottle
500 mL
≤5 EU/mL
≤30 mg/dL
Gibco™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers