missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, E.U.-approved, South America Origin

Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C. results.

Brand:  Gibco™ 10500056

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11543407

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Sera Category: Standard
Origin: E.U. Approved (South American)
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C. results.
Specifications

Specifications

Basic cell culture, specialty research, and specific assays
Store in freezer (–5 to –30°C)
E.U. Approved (South American)
Bottle
100 mL
Qualified
Liquid
Heat-Inactivated
EU-Approved
Foetal Bovine Serum (Heat Inactivated), E.U. Approved (South American Origin), 100mL, Liquid, –5 to –30°C Storage Temperature, Cattle/Bovine Species, Virus and Mycoplasma Tested, Dry Ice
Liquid
FBS Replacement Serum
Cattle/Bovine
Virus, Mycoplasma
Basic cell culture, specialty research, and specific assays
Qualified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers