Learn More
Cole-Parmer™ Fiberglass Insulated Type-K Thermocouple High-Temperature Wire Probes
Use this fiberglass insulated wire probe for high-temperature measurements. Cole-Parmer™ Fiberglass Insulated Type-K Thermocouple High-Temperature Wire Probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors.
Brand: Cole-Parmer™ 08467-24
Code : A0
Additional Details : Weight : 0.05000kg
Description
- High-Temperature Fiberglass Insulated Wire
- Response Time: 0.75 seconds or greater
- Custom designs or changes to this product are available upon request
- Fiberglass wire is not recommended for use in moist or wet environments
- Exposed-junction probes are not recommended for use with aqueous solutions
- Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
704°C | |
-150°C | |
-150°C to +704°C |