missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined

Product Code. 15498335
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498335 1000 g 1000g
15408345 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498335 Supplier Thermo Scientific Alfa Aesar Supplier No. H33152.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol

Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68855-54-9
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.988
MDL Number MFCD00132803
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
PubChem CID 10340
ChEBI CHEBI:29377
IUPAC Name disodium;carbonate
SMILES C(=O)([O-])[O-].[Na+].[Na+]

Specifications

Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Celite™ 545, treated with sodium carbonate, flux calcined

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.