Learn More
Filter aid, Celite Standard Super-cel
Description
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.
Solubility
Insoluble in water.
Notes
Incompatible with acids.
Specifications
Specifications
| Odor | Odorless |
| Quantity | 2500 g |
| CAS | 91053-39-3 |
| Merck Index | 14,4973 |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Filter aid, Celite Standard Super-cel |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.