Learn More
Hettich Lab Technology™ Filter Cards
Hettich Lab Technology™ Filter Cards are used for collecting cyto centrifuge samples and come in sizes to fit accommodate a variety of chambers and carriers.
POA
Specifications
Type Filter Card
View More Specs
Description
Description
  • For cyto centrifuge systems and cell evaluations
  • Suitable for a variety of cyto chambers and slide carriers
Specifications
Specifications
Filter Card