Learn More
Agilent BioTek Filter Cube
For use with Synergy LX
992.00 €
Product Code | Brand | Excitation/Emission | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Excitation/Emission | Price | Quantity & Availability | |||||
15998550
|
Agilent BioTek
1505006 |
EX 360/40, EM 460/40, 400 nm mirror (Blue) |
992.00 €
1 unit |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988550
|
Agilent BioTek
1505005 |
EX 485/20, EM 528/20, 510 nm mirror (Green) |
992.00 €
1 unit |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978550
|
Agilent BioTek
1505004 |
EX 530/25, EM 590/25, 570 nm mirror (Red) |
992.00 €
1 unit |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Synergy LX |