man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Filters and Filtration
Filter Plates
12010432
Print
Robu™ Filter Disc
Brand:
Robu™ 15403
Additional Details :
Weight :
0.01000kg
Product Code.
12010432
60.00 €
/ Pack of 10
Quantity
Estimated Shipment: 11-06-2024
Log in
to see stock.
Add to basket
Description
Product Suggestions
Videos
SDS
Documents
Certificates
Special Offers
Description
Description
40mm, porosity 2, thickness 4.5μm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers