missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Filter emission for darkroom 465nm to 495nm UVP
Brand: UVP 38-0361-01
Code : 04
Additional Details : Weight : 0.20000kg
Brand: UVP 38-0361-01
Code : 04
Additional Details : Weight : 0.20000kg