missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Filter emission for darkroom 668nm to 722nm UVP
Brand: UVP 38-0339-01
Code : 04
Additional Details : Weight : 0.20000kg
Brand: UVP 38-0339-01
Code : 04
Additional Details : Weight : 0.20000kg