missing translation for 'onlineSavingsMsg'
Learn More

UVP Filter emission for darkroom 700nm to 740nm UVP

Brand:  UVP 38-0369-01

Code : 04

Additional Details : Weight : 0.20000kg

Product Code. 12843808

  • 1670.00 € / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket