missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP Filter emission for darkroom 767nm to 807nm UVP
Brand: UVP 38-0362-01
Code : 04
Additional Details : Weight : 0.20000kg
Brand: UVP 38-0362-01
Code : 04
Additional Details : Weight : 0.20000kg