Learn More
ROBU VitraPOR™ Filter Funnel
Filter funnel available in a buchner shape or a conical shape.
50.65 € - 426.00 €
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Pore Size | Price | Quantity & Availability | ||||
10347921
|
ROBU VitraPOR™
21110 |
1000 mL | 0 |
150.00 €
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
10351597
|
ROBU VitraPOR™
21551 |
500 mL | 1 |
90.90 €
Each |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||||
16813046
|
ROBU VitraPOR™
22251 |
25 mL | 1 |
255.00 €
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16853046
|
ROBU VitraPOR™
22255 |
25 mL | 5 |
426.00 €
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16893026
|
ROBU VitraPOR™
2111F |
1000 mL | Fine |
213.00 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16803036
|
ROBU VitraPOR™
2112F |
125 mL | Fine |
69.90 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16813036
|
ROBU VitraPOR™
21250 |
250 mL | 0 |
75.00 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16823036
|
ROBU VitraPOR™
21255 |
250 mL | 5 |
129.00 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16833036
|
ROBU VitraPOR™
2125F |
250 mL | Fine |
103.00 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16843036
|
ROBU VitraPOR™
2150F |
50 mL | Fine |
50.65 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16853036
|
ROBU VitraPOR™
2155F |
500 mL | Fine |
128.00 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
16863036
|
ROBU VitraPOR™
2175F |
75 mL | Fine |
54.70 €
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass 3.3 |