missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Filter Papers, School Grade

General purpose cotton filter papers (12.5cm Dia) for use in classrooms for educational purposes.

Brand:  Macherey-Nagel™ 4007130125

Code : 02

Additional Details : Weight : 0.60000kg

Product Code. 12962701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

General purpose filtration
Schools grade
100 Pack
125 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers