Learn More
Eppendorf™ Filter Set for BioSpectrometer Fluorescence
Brand:  Eppendorf™ 6137928009
Code : 99
Additional Details : Weight : 0.50000kg
Product Code. 15139552
  • 3780.00 € / Each
Estimated Shipment: 26-04-2024
to see stock.
Description
Description
Order Info
ForUseWithEquipment
Specifications
Specifications
Filter Set
For verifying photometric accuracy and wavelength systematic error (according to NIST) and verifying fluorimetric precision (random error) and linearity
For BioSpectrometer fluorescence