Learn More
Sartorius™ Complete Filter Support
Conveniently designed with a centering rim for easy positioning under the funnel. Sartorius™ Complete Filter Support prevents the inserted membrane filter from being displaced in an all-glass vacuum filter assembly.
Brand: Sartorius™ 6983004
Code : NEW
Additional Details : Weight : 0.09000kg
Description
Conveniently designed with a centering rim for easy positioning under the funnel. Sartorius™ Complete Filter Support prevents the inserted membrane filter from being displaced in an all-glass vacuum filter assembly.
- Centering rim
- Includes glass frit, PTFE ring, fluoroelastomer O-ring
- For use with all-glass vacuum filter assembly
Specifications
Sartorius™ All-Glass Vacuum Filter Holder system | |
Glass frit, PTFE ring, fluoroestamoer O-ring | |
1/ea. | |
Complete Filter Support |