missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Fine Tip Micro-Pasteur Pipette, Bulk Pack

Product Code. 16454999
Click to view available options
:
Packaging: Bulk Peel Pouch
This item is not returnable. View return policy

Product Code. 16454999

Brand: Wheaton™ 711117MF/3000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pasteur pipette for simple and safe liquid measurement and transfer.

  • Ensures safe sample transfer
  • Micro size
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bulb Draw (Metric) 1.5 mL
Calibrated No
Certifications/Compliance CE Marked
Description Fine Tip Plastic Micro Pasteur Pipette
Material LDPE
Sterility Sterile
For Use With (Application) Liquid transfer
Graduated No
Quantity 3000/Pk
Tip Style Fine Tip
Top Style Bulb
Packaging Type Bulk Peel Pouch
Length (Metric) 150 mm
Disposable Single-use
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.