missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finntip™ Flex Filtered Pipette Tips
GREENER_CHOICE

Product Code. 11802713
Change view
Click to view available options
Volume (Metric):
10 μL
30 μL
100 μL
200 μL
300 μL
1000 μL
1200 μL
10 mL
Unit Size:
Pack of 50
Pack of 960
Product Code. Volume (Metric) unitSize
11802713 1000 μL Pack of 960
11833430 10 μL Pack of 960
11843430 30 μL Pack of 960
11853430 100 μL Pack of 960
11863430 200 μL Pack of 960
11852703 300 μL Pack of 960
11862703 1200 μL Pack of 960
11475558 10 mL Pack of 50
8 options
This item is not returnable. View return policy

Product Code. 11802713

Brand: Thermo Scientific™ 94056710

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Minimize the risk of cross-contamination with these flex filtered pipette tips that combine the ergonomics of a standard pipette tip with the safety of a filter tip.

Minimize the risk of cross-contamination with Thermo Scientific™ Finntip™ Flex Filter Pipette Tips, which combine the ergonomics of a standard Finntip Flex pipette tip and the safety of a filter tip. Offered sterile, and free from DNA, DNase and RNase. Ideal for PCR and pipetting of radioactive and infectious samples. The filters, made of an inert nonsealing polyethylene matrix, are effective at eliminating carryover contamination from liquids and aerosols between pipetter and tip.

Ordering Information:

Supplied in vacuum-sealed sterilized tip racks.

Recommended for:

Use in sensitive applications such as PCR and pipetting of radioactive and infectious samples.

TRUSTED_SUSTAINABILITY

Specifications

Description Finntip Flex Filter 1000 μL, sterile
Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Length (Metric) 8.7 cm
Format Rack
Color Code Blue
For Use With (Equipment) Thermo Scientific Finnpipette
Product Line FinnTip Flex
Fit for Purpose Sterile & PCR Ready
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.