Learn More
Integra Biosciences™ Gas Cartridge Adapter, FIREBOY™ CP250+
Use this adapter to fit Campingaz CP 250+ gas cartridges to your FIREBOY™ portable safety bunsen burner.
Brand: Integra Biosciences™ 144051
Code : 04
Additional Details : Weight : 0.90000kg
Description
- Suitable for FIREBOY™ portable safety bunsen burner
- For Campingaz CP 25+ cartridges
Order Info
ForUseWithEquipment
Specifications
Adapter | |
Campingaz CP 250+ 250g Butane Cartridge, FIREBOY Portable Safety Bunsen Burner |