Learn More
Fisher BioReagents™ Bovine Serum Albumin (BSA) Low Endotoxin Powder
Purified by a heat shock process with additional treatments
Brand: Fisher BioReagents™ BP9705-100
Code : A0
Additional Details : CAS Number : 9048-46-8 Weight : 0.10000kg
Description
CAS: 9048-46-8
Specifications
BSA Low Endotoxin | |
Proprietary heat shock process | |
Bovine |
6.5 to 7.5 | |
100g | |
Powder |
Safety and Handling
- Bovine Serum Albumin, Heat Shock, Reagent Grade Powder, pH 7.0
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.