Learn More
Fisher BioReagents™ Bovine Serum Albumin (BSA) Microbiological Grade Powder
Purified by a modified cold ethanol procedure
Brand: Fisher BioReagents™ BP9700-100
Code : NEW
Additional Details : CAS Number : 9048-46-8 Weight : 0.79380kg
Description
CAS: 9048-46-8
Specifications
BSA Microbiological Grade | |
Modified cold ethanol procedure | |
Bovine |
6.8 to 7.2 | |
100g | |
Powder |
Safety and Handling
- Bovine Serum Albumin, Heat Shock, Reagent Grade Powder, pH 7.0
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.